Caprylsäure
Bottle
Katalognr. APOSOR6008-25G
New Product
Anbieter: Apollo Scientific
Direkt zu
Product Details & Documents
Synonyme: Octanoic acid, n-Octanoic Acid, Octoic acid
- CAS Nummer:124-07-2
- Formel:C₈H₁₆O₂
- MDL-Nummer:MFCD00004429
- Molekulargewicht:144.21 g/mol
- Siedepunkt:236…238 °C
- Schmelzender Platingehalt:15…17 °C
- Dichte:0,91 g/cm³ (25 °C)
- Blitz Pt:>110 °C
- Lagertemperatur:Raumtemperatur
- EINECS:204-677-5
- UN:3265
- ADR:8,III
Technische Daten
Technische Daten
- Packung:Bottle
Technische Daten
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



