Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
 

4'-Cyclohexylacetophenon 98%

Bottle
Catalog # APOSOR4954-2.5KG
New Product
Supplier:   Apollo Scientific
4'-Cyclohexylacetophenon 98%
Catalog # APOSOR4954-2.5KG
Supplier:  Apollo Scientific
New Product
CAS Number:  18594-05-3
4'-Cyclohexylacetophenon 98%

Bottle

CAS Number:  18594-05-3
Price
SFr. 5’340.00
Log in for availability
1 * 2,5 kg
Product Details & Documents
Synonyms:  p-Cyclohexylacetophenone
  • CAS Number:18594-05-3
  • Formula:C₁₄H₁₈O
  • MDL Number:MFCD00001453
  • Molecular Weight:202.3 g/mol
  • Boiling Pt:179…181 °C/14mmHg
  • Melting Pt:66…70 °C
  • Storage Temperature:Raumtemperatur
Specifications

Specifications

  • Packung:
    Bottle

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.