Tallowamine polyethoxylated
Tallowamine polyethoxylated
EHERC17136000
- Packing group: III
- Bezeichnung:Tallowamine polyethoxylated
- UN No.:3082
- Cat. No.:EHERC17136000
- CAS Nr.:61791-26-2
- GHS-Signal:Achtung
- Beschreibung Gefahrenklasse:GHS05, GHS07, GHS09
- Lagerbedingungen:20 °C
- Synonym(s):Agnique TAM 15;Alkaminox T 12;Amiet 502;Amiladin D;Armostat 300;Atlas G 3762;Atmer 261;Berol 387;Chemeen T 5;Cirrasol G 3780A;Crisomin T 15;Crodamet T 15;Emulson AG/GPE 3CA;Ethomeen T 105;Ethomeen T 12E;Ethomeen YT;Ethox TAM 20;Ethoxamine SF 11;Ethoxylated Tallow Alkyl Amines;Ethylan TT 203;Fentacare T 15;Genamin 267;Genamin T 020;Genamin T 200;Imbentin TAM 400;Hyspray;Icomeen T 15;Imbentin TAM 400;Katapol PN 430;Kemamine AS 974-1;Mazeen T 2;Naylamul S 11;Nimine T 2-210 ;Nissan Nymeen T 2-202;Noramox S;Noramox S 11;Nymeen T 2-202;PEG 100 tallow amine;Rhodameen IT 50/46;Surfonic AGM 510;Surfonic T 15;TAM 105;Teric 17M2;Terwet 3780;Toximul TA 5;Trymeen 6606;VP 532SPB;Varonic T 205;Weedmaster CT Surfactant;Witcamine TAM 105
- Shipping conditions:RT
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



