Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
Amidosulfuron

Amidosulfuron

Amidosulfuron
Catalog # EHERC10162000
Supplier:   EHRENSTORFER
Amidosulfuron
Catalog # EHERC10162000
Supplier:  EHRENSTORFER
CAS Number:  
Amidosulfuron
Amidosulfuron

Amidosulfuron

CAS Number:  
Price
SFr. 304.00
Log in for availability
1 * 0,1 g
Product Details & Documents
  • SMILES: COc1cc(OC)nc(NC(=O)NS(=O)(=O)N(C)S(=O)(=O)C)n1
  • Inchi: InChI=1S/C9H15N5O7S2/c1-14(22(4,16)17)23(18,19)13-9(15)12-8-10-6(20-2)5-7(11-8)21-3/h5H,1-4H3,(H2,10,11,12,13,15)
  • IUPAC: 1-(4,6-dimethoxypyrimidin-2-yl)-3-[methyl(methylsulfonyl)sulfamoyl]urea
  • Packing group: III
Specifications

Specifications

  • Bezeichnung:
    Amidosulfuron
  • UN No.:
    3077
  • Cat. No.:
    EHERC10162000
  • CAS Nr.:
    120923-37-7
  • GHS-Signal:
    Achtung
  • Molekulargewicht:
    369,37
  • Beschreibung Gefahrenklasse:
    GHS07, GHS09
  • Lagerbedingungen:
    20 °C
  • Synonym(s):
    3,5-Dithia-2,4-diazahexanamide, N-(4,6-dimethoxy-2-pyrimidinyl)-4-methyl-, 3,3,5,5-tetraoxide;2,4-Dithia-3,5-diazahexan-6-amide, N-(4,6-dimethoxy-2-pyrimidinyl)-3-methyl-, 2,2,4,4-tetraoxide (9CI);Adret;Amidosulfuron;Cratil;Gratil;Grodyl;HOE 075032;Hoestar
  • Shipping conditions:
    RT
  • Formel:
    C₉H₁₅N₅O₇S₂

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.