Hexylresorcinol
Hexylresorcinol
EHERC14209200
- SMILES: CCCCCCc1ccc(O)cc1O Inchi: InChI=1S/C12H18O2/c1-2-3-4-5-6-10-7-8-11(13)9-12(10)14/h7-9,13-14H,2-6H2,1H3 IUPAC: 4-hexylbenzene-1,3-diol
- Packing group: III
- Bezeichnung:Hexylresorcinol
- UN No.:3077
- Cat. No.:EHERC14209200
- CAS Nr.:136-77-6
- GHS-Signal:Achtung
- Molekulargewicht:194,27
- Beschreibung Gefahrenklasse:GHS07, GHS09
- Lagerbedingungen:20 ℃
- Synonym(s):Hexylresorcinol; 4-Hexyl-1,3-benzenediol; 4-Hexylresorcinol; 1,3-Dihydroxy-4-hexylbenzene; 1-(2',4'-Dihydroxyphenyl)hexane; 4-(1-Hexyl)resorcinol; 4-Hexyl-1,3-benzenediol; 4-Hexyl-1,3-dihydroxybenzene; 4-Hexylresorcine; 4-Hexylresorcinol; 4-n-Hexylresorcinol; Adrover; Antascarin; Ascaricid; Ascarinol; Ascaryl; Caprokol; Crystoids; Cystoids Anthelmintic; EverFresh; Gelovermin; Hexylresorcinol; Hidesol; KOM 14; Mycoderm; NSC 1570; Oxana; Prensol; S.T. 37; Sucrets; Worm-Agen; p-Hexylresorcinol
- Shipping conditions:RT
- Formel:C₁₂H₁₈O₂



