Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
Chloroxuron

Chloroxuron

Chloroxuron
Catalog # EHERC11540000
Supplier:   EHRENSTORFER
Chloroxuron
Catalog # EHERC11540000
Supplier:  EHRENSTORFER
CAS Number:  
Chloroxuron
Chloroxuron

Chloroxuron

CAS Number:  
Price
SFr. 112.00
Log in for availability
1 * 0,25 g
Product Details & Documents
  • SMILES: CN(C)C(=O)Nc1ccc(Oc2ccc(Cl)cc2)cc1
  • Inchi: InChI=1S/C15H15ClN2O2/c1-18(2)15(19)17-12-5-9-14(10-6-12)20-13-7-3-11(16)4-8-13/h3-10H,1-2H3,(H,17,19)
  • IUPAC: 3-[4-(4-chlorophenoxy)phenyl]-1,1-dimethylurea
  • Packing group: III
Specifications

Specifications

  • Bezeichnung:
    Chloroxuron
  • UN No.:
    3077
  • Cat. No.:
    EHERC11540000
  • CAS Nr.:
    1982-47-4
  • GHS-Signal:
    Achtung
  • Molekulargewicht:
    290,74
  • Beschreibung Gefahrenklasse:
    GHS07, GHS09
  • Lagerbedingungen:
    20 °C
  • Synonym(s):
    Urea, 3-[p-(p-chlorophenoxy)phenyl]-1,1-dimethyl- (7CI,8CI); N'-[4-(4-Chlorophenoxy)phenyl]-N,N-dimethylurea; 3-[4-(p-Chlorophenoxy)phenyl]-1,1-dimethylurea; 3-[p-(p-Chlorophenoxy)phenyl]-1,1-dimethylurea; C 1983; Chloroxifenidim; Chloroxuron; Gesamoos; N-4-(p-Chlorophenoxy)phenyl-N',N'-dimethyl urea; N1-[4-(4-Chlorophenoxy)phenyl]-N,N-dimethylurea; Norex; Tenoran
  • Shipping conditions:
    RT
  • Formel:
    C₁₅H₁₅ClN₂O₂

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.