Acephate
Acephate
Katalognr. EHERC10010000
Anbieter: EHRENSTORFER
Direkt zu
Product Details & Documents
- SMILES: COP(=O)(NC(=O)C)SC
- Inchi: InChI=1S/C4H10NO3PS/c1-4(6)5-9(7,8-2)10-3/h1-3H3,(H,5,6,7)
- IUPAC: N-[methoxy(methylsulfanyl)phosphoryl]acetamide
- Packing group: III
Technische Daten
Technische Daten
- Bezeichnung:Acephate
- UN No.:3077
- Cat. No.:EHERC10010000
- CAS Nr.:30560-19-1
- GHS-Signal:Achtung
- Molekulargewicht:183,17
- Beschreibung Gefahrenklasse:GHS07, GHS09
- Lagerbedingungen:–18 °C
- Synonym(s):Phosphoramidothioic acid, N-acetyl-, O,S-dimethyl ester (ACI); Phosphoramidothioic acid, acetyl-, O,S-dimethyl ester (8CI, 9CI); Acefate; Acetamidophos; Aciphid; Ant Hayter; Asataf; DMAP; Fire Ant Hayter Bait; Lepitect; O,S-Dimethyl acetylphosphoramidothioate; O,S-Dimethyl N-acetylphosphoramidothioate; O,S-Dimethyl N-acetylphosphoramidothiolate; Orthene; Orthene 75SP; Orthene 97; Orthene PCO; Orthene S; Orthene TTO; Orthene TTO 97; Orthene-755; Orthorane; Ortran; RE 12420
- Shipping conditions:RT
- Formel:C₄H₁₀NO₃PS
Technische Daten
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



