Benzalkonium chloride 80%
New Product
Anbieter: Apollo Scientific
Direkt zu
Über diesen Artikel
Usage: Organic building block/reagent
Synonyme: Quaternary ammonium compounds, benzyl-C8-18-alkyldimethyl, chlorides, Alkylbenzyldimethylammonium chloride
- CAS Nummer:68424-85-1
- MDL-Nummer:MFCD00145757
- EINECS:270-325-2
- UN:1760
6 Varianten
Product Details & Documents
Usage: Organic building block/reagent
Synonyme: Quaternary ammonium compounds, benzyl-C8-18-alkyldimethyl, chlorides, Alkylbenzyldimethylammonium chloride
- CAS Nummer:68424-85-1
- MDL-Nummer:MFCD00145757
- EINECS:270-325-2
- UN:1760
Product Family Options
Produktinformationen
- PackungVerfügbarkeitPreis
- Spezifikationen:New ProductNew Product
- Spezifikationen:New ProductNew Product
- Spezifikationen:New ProductNew Product
- Spezifikationen:New ProductNew Product
- Spezifikationen:New ProductNew Product
- Spezifikationen:New ProductNew Product
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



