AuftragsverfolgungKontakt
Docosanoic acid 85%
New Product
Docosanoic acid 85%
New Product
Docosanoic acid 85%
Usage: Organic building block/reagent
 :  Docosanoic acid
  •  :112-85-6
  •  :C₂₂H₄₄O₂
  •  :MFCD00002807
  •  :340,58 g/mol
Product Family Options
  • Packung
  • Bottle
    1 * 25 g
    1 * 25 g - SFr. 26.00
    SFr. 26.00
    1 * 25 g
       SFr. 26.00
    New Product
    New Product
  • Bottle
    1 * 100 g
    1 * 100 g - SFr. 29.50
    SFr. 29.50
    1 * 100 g
       SFr. 29.50
    New Product
    New Product
  • Bottle
    1 * 500 g
    1 * 500 g - SFr. 69.30
    SFr. 69.30
    1 * 500 g
       SFr. 69.30
    New Product
    New Product
  • Bottle
    1 * 2,5 kg
    1 * 2,5 kg - SFr. 304.00
    SFr. 304.00
    1 * 2,5 kg
       SFr. 304.00
    New Product
    New Product