Docosanoic acid 85%
New Product
Usage: Organic building block/reagent
: Docosanoic acid
- :112-85-6
- :C₂₂H₄₄O₂
- :MFCD00002807
- :340,58 g/mol
Usage: Organic building block/reagent
: Docosanoic acid
- :112-85-6
- :C₂₂H₄₄O₂
- :MFCD00002807
- :340,58 g/mol
Product Family Options
- Packung
- New ProductNew Product
- New ProductNew Product
- New ProductNew Product
- New ProductNew Product