AuftragsverfolgungKontakt
Docosanoic acid ≥98%
New Product
Docosanoic acid ≥98%
New Product
Docosanoic acid ≥98%
Usage: Biochemical/life science applications
 :  Docosanoic acid
  •  :112-85-6
  •  :C₂₂H₄₄O₂
  •  :340,58 g/mol
Product Family Options
  • Packung
  • Bottle
    1 * 500 mg
    1 * 500 mg - SFr. 74.50
    SFr. 74.50
    1 * 500 mg
       SFr. 74.50
    New Product
    New Product
  • Bottle
    1 * 1 g
    1 * 1 g - SFr. 108.00
    SFr. 108.00
    1 * 1 g
       SFr. 108.00
    New Product
    New Product