Bromophenol Red Indicator biologische Färbemittel
Usage: Organic building block/ reagent.
: 4,4'-(3H-2,1-Benzoxathiol-3-ylidene)bis[2-bromophenol] S,S-dioxide
- :2800-80-8
- :C₁₉H₁₂Br₂O₅S
- :MFCD00023014
- :512,17 g/mol
Usage: Organic building block/ reagent.
: 4,4'-(3H-2,1-Benzoxathiol-3-ylidene)bis[2-bromophenol] S,S-dioxide
- :2800-80-8
- :C₁₉H₁₂Br₂O₅S
- :MFCD00023014
- :512,17 g/mol
Product Family Options
- Packung
- New ProductNew Product
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.