AuftragsverfolgungKontakt
Torkinib ≥98%
New Product
Torkinib ≥98%
New Product
Torkinib ≥98%
Usage: Biochemical/ life science applications.
 :  2-(4-Amino-1-isopropyl-1H-pyrazolo[3,4-d]pyrimidin-3-yl)-1H-indol-5-ol, Torkinib, 2-[4-Amino-1-(1-methylethyl)-1H-pyrazolo[3,4-d]pyrimidin-3-yl]-1H-indol-5-ol
  •  :1092351-67-1
  •  :C₁₆H₁₆N₆O
  •  :308,34 g/mol
Product Family Options
  • Packung
  • Bottle
    1 * 50 mg
    1 * 50 mg - SFr. 212.00
    SFr. 212.00
    1 * 50 mg
       SFr. 212.00
    New Product
    New Product
  • Bottle
    1 * 100 mg
    1 * 100 mg - SFr. 364.00
    SFr. 364.00
    1 * 100 mg
       SFr. 364.00
    New Product
    New Product
  • Bottle
    1 * 200 mg
    1 * 200 mg - SFr. 541.00
    SFr. 541.00
    1 * 200 mg
       SFr. 541.00
    New Product
    New Product