Biotin ≥98%
New Product
Anbieter: Apollo Scientific
Direkt zu
Über diesen Artikel
Usage: Biochemical/ life science applications.
- Store at 2 to 8 °C Non-hazardous
Synonyme: Vitamin H, D-Biotin, Vitamin B8, 5-((3aS,4S,6aR)-2-Oxohexahydro-1H-thieno[3,4-d]imidazol-4-yl)pentanoic acid, Vitamin B7
- CAS Nummer:58-85-5
- Formel:C₁₀H₁₆N₂O₃S
- Molekulargewicht:244,31 g/mol
- Lagertemperatur:Kühlschrank
- EINECS:200-399-3
3 Varianten
Product Details & Documents
Usage: Biochemical/ life science applications.
- Store at 2 to 8 °C Non-hazardous
Synonyme: Vitamin H, D-Biotin, Vitamin B8, 5-((3aS,4S,6aR)-2-Oxohexahydro-1H-thieno[3,4-d]imidazol-4-yl)pentanoic acid, Vitamin B7
- CAS Nummer:58-85-5
- Formel:C₁₀H₁₆N₂O₃S
- Molekulargewicht:244,31 g/mol
- Lagertemperatur:Kühlschrank
- EINECS:200-399-3
Product Family Options
Produktinformationen
- PackungVerfügbarkeitPreis
- Spezifikationen:New ProductNew Product
- Spezifikationen:New ProductNew Product
- Spezifikationen:New ProductNew Product
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



