Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
 

Shield-1 ≥98%

New Product
Supplier:   Apollo Scientific
Shield-1 ≥98%
Supplier:  Apollo Scientific
New Product
Certificates

4 Options Available Below

Product Details & Documents

Usage: Biochemical/ life science applications.

Synonyms:  Shield-1
  • CAS Number:914805-33-7
  • Formula:C₄₂H₅₆N₂O₁₀
  • Molecular Weight:748,9 g/mol
Product Family Options

Product Information

  • Packung
    Availability
    Price
  • Bottle
    Log in for availability
    Log in for availability
    1 * 25 mg
    1 * 25 mg - SFr. 1’440.00
    SFr. 1’440.00
    1 * 25 mg
       SFr. 1’440.00

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 50 mg
    1 * 50 mg - SFr. 1’950.00
    SFr. 1’950.00
    1 * 50 mg
       SFr. 1’950.00

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 100 mg
    1 * 100 mg - SFr. 2’620.00
    SFr. 2’620.00
    1 * 100 mg
       SFr. 2’620.00

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 500 mg
    1 * 500 mg - SFr. 5’220.00
    SFr. 5’220.00
    1 * 500 mg
       SFr. 5’220.00

    Specifications:
    New Product
    New Product