Methyl Linoleate ≥98%
Bottle
Katalognr. APOSBUP18978-1G
New Product
Anbieter: Apollo Scientific
Methyl Linoleate ≥98%
Katalognr. APOSBUP18978-1G
Anbieter: Apollo Scientific
New Product
CAS Nummer: 112-63-0
Direkt zu
Product Details & Documents
Usage: Biochemical/life science applications.
Synonyme: cis,cis-9,12-Octadecadienoic acid methyl ester, 9,12-Octadecanoic acid methyl ester, 9Z,12Z-Octadecadienoic acid methyl ester, Linoleic acid methyl ester, (9Z,12Z)-Methyl octadeca-9,12-dienoate, Methyl cis,cis-9,12-octadecadienoate, 9Z,12Z-Oktadekadiensäuremethylester, 9,12 Methyl octadecaotenoate methyl linoleate, cis,cis-9,12-Oktadekadiensäuremethylester
- CAS Nummer:112-63-0
- Formel:C₁₉H₃₄O₂
- Molekulargewicht:294,47 g/mol
Technische Daten
Technische Daten
- Packung:Bottle



