Methyl Tridecanoate ≥95%
Bottle
Katalognr. APOSBUP18942-1G
New Product
Anbieter: Apollo Scientific
Methyl Tridecanoate ≥95%
Katalognr. APOSBUP18942-1G
Anbieter: Apollo Scientific
New Product
CAS Nummer: 1731-88-0
Direkt zu
Product Details & Documents
Usage: Biochemical/life science applications.
Synonyme: Tridecylic acid methyl ester, Tridecanoic acid methyl ester
- CAS Nummer:1731-88-0
- Formel:C₁₄H₂₈O₂
- Molekulargewicht:228,37 g/mol
Technische Daten
Technische Daten
- Packung:Bottle



