Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
 

Heptadecanoic acid ≥98%

New Product
Supplier:   Apollo Scientific
Heptadecanoic acid ≥98%
Supplier:  Apollo Scientific
New Product
Certificates
Heptadecanoic acid ≥98%

2 Options Available Below

Product Details & Documents

Usage: Biochemical/life science applications.

Synonyms:  Heptadecanoic acid, Heptadecylic acid
  • CAS Number:506-12-7
  • Formula:C₁₇H₃₄O₂
  • Molecular Weight:270,45 g/mol
Product Family Options

Product Information

  • Packung
    Availability
    Price
  • Bottle
    Log in for availability
    Log in for availability
    1 * 500 mg
    1 * 500 mg - SFr. 48.50
    SFr. 48.50
    1 * 500 mg
       SFr. 48.50

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - SFr. 67.60
    SFr. 67.60
    1 * 1 g
       SFr. 67.60

    Specifications:
    New Product
    New Product