AuftragsverfolgungKontakt
Dynamic Ovens for Rapid Temperature Changes, MK | Avantor
Dynamic Ovens for Rapid Temperature Changes, MK
New Product
Anbieter:   Binder
Dynamic Ovens for Rapid Temperature Changes, MK
Anbieter:  Binder
New Product
Zertifikate
Dynamic Ovens for Rapid Temperature Changes, MK
3 Varianten
Product Details & Documents
The BINDER MK series chambers are suitable for all heat and cold tests between –40 and +180 ℃. The MK is a clever alternative to complex individual solutions for cyclical temperature tests.
  • Programmable condensation protection for test material
  • Heated viewing window with LED interior lighting
  • Intuitive touchscreen controller with time-segment and real-time programming
  • APT.line™ preheating chamber technology
  • Class 2 independent adjustable temperature safety device (DIN 12880) with visual alarm
  • Internal data logger, measured values can be read out in open format via USB
  • Troubleshooting system with visual and audible alarms
  • CFC-free refrigerant R-452A
  • Adjustable ramp function, real-time clock and door heating
Units have an inner chamber made of stainless steel, they feature cooling with a compressor cooling unit, an integrated chart recorder,  4 casters, two with brakes and an Ethernet interface. The 240 and 400 models have a 50 mm access port with silicone plug on the left. The larger 720 models have 2×80 mm access ports with silicone plugs on the left and right.
The use of CO₂, a natural refrigerant with a GWP value = 1, meets the highest environmental standards and helps to reduce operating costs on a long-term basis. The robust system quality, along with the extensive basic equipment and maintenance-friendly design ensure reliable operation during continuous use.
Lieferinformationen: Supplied with one stainless steel rack, Multi Management Software APT-COM™ Basic Edition and Binder test certificate. Calibrations and validations possible. A wide range of options and accessories are available, including compressed air supply system, access ports. For details please contact your local Avantor sales office.
Product Family Options
Produktinformationen
  • Bezeichnung
    Verfügbarkeit
    Preis
  • Dynamic climate chamber MK 240 CO₂
    Einloggen für Verfügbarkeit
    Einloggen für Verfügbarkeit
    1 * 1 ST
    1 * 1 ST
    Spezifikationen:
    New Product
    New Product
  • Einloggen für Verfügbarkeit
    Einloggen für Verfügbarkeit
    1 * 1 ST
    1 * 1 ST
    Spezifikationen:
    New Product
    New Product
  • Einloggen für Verfügbarkeit
    Einloggen für Verfügbarkeit
    1 * 1 ST
    1 * 1 ST
    Spezifikationen:
    New Product
    New Product