DAPT ≥98%
New Product
Anbieter: Apollo Scientific
Direkt zu
Über diesen Artikel
Usage: Biochemical/life science applications
Synonyme: N-[N-(3,5-Difluorophenacetyl-L-alanyl)]-S-phenylglycine t-Butyl Ester, tert-Butyl (S)-2-((S)-2-(2-(3,5-difluorophenyl)acetamido)propanamido)-2-phenylacetate, (3,5-Difluorophenylacetyl)-Ala-Phg-OtBu, (3,5-Difluorophenylacetyl)-L-alanyl-L-2-phenylglycine tert-Butyl Ester
- CAS Nummer:208255-80-5
- Formel:C₂₃H₂₆F₂N₂O₄
- Molekulargewicht:432,46 g/mol
3 Varianten
Product Details & Documents
Usage: Biochemical/life science applications
Synonyme: N-[N-(3,5-Difluorophenacetyl-L-alanyl)]-S-phenylglycine t-Butyl Ester, tert-Butyl (S)-2-((S)-2-(2-(3,5-difluorophenyl)acetamido)propanamido)-2-phenylacetate, (3,5-Difluorophenylacetyl)-Ala-Phg-OtBu, (3,5-Difluorophenylacetyl)-L-alanyl-L-2-phenylglycine tert-Butyl Ester
- CAS Nummer:208255-80-5
- Formel:C₂₃H₂₆F₂N₂O₄
- Molekulargewicht:432,46 g/mol
Product Family Options
Produktinformationen
- PackungVerfügbarkeitPreis
- Spezifikationen:New ProductNew Product
- Spezifikationen:New ProductNew Product
- Spezifikationen:New ProductNew Product
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



