Ampicillin Trihydrate ≥98%
Bottle
Katalognr. APOSBUP18462-1G
New Product
Anbieter: Apollo Scientific
Ampicillin Trihydrate ≥98%
Katalognr. APOSBUP18462-1G
Anbieter: Apollo Scientific
New Product
CAS Nummer: 7177-48-2
Direkt zu
Product Details & Documents
Usage: Biochemical/life science applications
Synonyme: (2S,5R,6R)-6-((R)-2-Amino-2-phenylacetamido)-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid trihydrate
- CAS Nummer:7177-48-2
- Formel:C₁₆H₁₉N₃O₄S·3H₂O
- Molekulargewicht:403,45 g/mol
Technische Daten
Technische Daten
- Packung:Bottle
Technische Daten
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



