Benzaldehyde 99+%
Anbieter: Apollo Scientific
Direkt zu
Über diesen Artikel
Usage: Organic building block/ reagent.
- CAS Nummer:100-52-7
- Formel:C₇H₆O
- MDL-Nummer:MFCD00003299
- Molekulargewicht:106,12194 g/mol
- EINECS:202-860-4
- UN:1990
2 Varianten
Product Details & Documents
Usage: Organic building block/ reagent.
- CAS Nummer:100-52-7
- Formel:C₇H₆O
- MDL-Nummer:MFCD00003299
- Molekulargewicht:106,12194 g/mol
- EINECS:202-860-4
- UN:1990



