Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
Benzoic acid ≥98%

Benzoic acid ≥98%

Bottle
Catalog # APOSBUP19181-1G
New Product
Supplier:   Apollo Scientific
Benzoic acid ≥98%
Catalog # APOSBUP19181-1G
Supplier:  Apollo Scientific
New Product
CAS Number:  65-85-0
Benzoic acid ≥98%

Bottle

CAS Number:  65-85-0
Price
45.10 SFr.
Log in for availability
1 * 1 g
Product Details & Documents

Usage: Biochemical/ life science applications.

  • CAS Number:65-85-0
  • Formula:C₇H₆O₂
  • Molecular Weight:122,12 g/mol
Specifications

Specifications

  • Packung:
    Bottle

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.