AuftragsverfolgungKontakt
Hydroxyamine hydrochloride ≥98%
New Product
Hydroxyamine hydrochloride ≥98%
New Product
Hydroxyamine hydrochloride ≥98%
Usage: Biochemical/ life science applications.
 :  Hydroxylamine HCl, Hydroxylamine hydrochloride
  •  :5470-11-1
  •  :ClH₄NO
  •  :69,49 g/mol
Product Family Options
  • Packung
  • Bottle
    1 * 500 mg
    1 * 500 mg - SFr. 48.50
    SFr. 48.50
    1 * 500 mg
       SFr. 48.50
    New Product
    New Product
  • Bottle
    1 * 1 g
    1 * 1 g - SFr. 67.60
    SFr. 67.60
    1 * 1 g
       SFr. 67.60
    New Product
    New Product
  • Bottle
    1 * 5 g
    1 * 5 g - SFr. 151.00
    SFr. 151.00
    1 * 5 g
       SFr. 151.00
    New Product
    New Product
  • Bottle
    1 * 10 g
    1 * 10 g - SFr. 226.00
    SFr. 226.00
    1 * 10 g
       SFr. 226.00
    New Product
    New Product