5-hydroxytryptophan ≥98%
New Product
Anbieter: Apollo Scientific
Direkt zu
Über diesen Artikel
Usage: Biochemical/ life science applications.
Synonyme: DL-5-Hydroxytryptophan, 2-Amino-3-(5-hydroxy-1H-indol-3-yl)propanoic acid
- CAS Nummer:56-69-9
- Formel:C₁₁H₁₂N₂O₃
- Molekulargewicht:220,22 g/mol
2 Varianten
Product Details & Documents
Usage: Biochemical/ life science applications.
Synonyme: DL-5-Hydroxytryptophan, 2-Amino-3-(5-hydroxy-1H-indol-3-yl)propanoic acid
- CAS Nummer:56-69-9
- Formel:C₁₁H₁₂N₂O₃
- Molekulargewicht:220,22 g/mol



