Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
 

High Magnification Microscope, Edge 3.0

Mikroskop mit hoher Vergrößerung, 700× - 900×, 8 LED Weiß, 5,0 Megapixel
Katalognr. IDCPAM73915MT8
New Product
Irregular Voltage
High Magnification Microscope, Edge 3.0
Katalognr. IDCPAM73915MT8
New Product
Irregular Voltage
High Magnification Microscope, Edge 3.0

Mikroskop mit hoher Vergrößerung, 700× - 900×, 8 LED Weiß, 5,0 Megapixel

Preis
SFr. 1’280.00
Einloggen für Verfügbarkeit
1 * 1 ST
Product Details & Documents

The Edge 3.0 microscope offers DPQ, EDOF, eFLC, AMR, EDR, a click-to-focus functionality for automatically changing focus to the point of interest, and smooth live imaging with a resolution of 5,0 MP at 15 fps or 1,3 MP at 45 fps. This microscope has a narrow working distance and requires the use of a stand with precision control to adjust focus.

  • Advanced CMOS image sensor allows to transmit sharp and crisp image

  • Extended Depth of Field (EDOF)
  • Extended Dynamic Range (EDR)
  • Automatic Magnification Reading (AMR)
  • Enhanced Flexible LED Control (eFLC)
  • 700 to 900× magnification

    Measurement and Calibration

    Microtouch sensor

It is a tool for biomedical and scientific research, material analysis, electronics inspection, or any similar applications that require high magnification, versatility, and mobility. Its USB 3,0 connection ensures fast data transfer and reliable performance.

Interface: USB 3,0

Operating system: Windows 7/8/10/11 macOS 10.14 or later; Windows 7, 8, 10 & 11, MacOS 10.12 and up

Supported image formats: BMP, GIF, PNG, MNG, TIF, TGA, PCX, WBMP, JP2, JPC, JPG, PGX, RAS, PNM (Windows); PNG, JPEG (MacOS)

Supported video formats: WMV, FLV, SWF (Windows); MOV (MacOS)

Housing material: Metal housing

Dimensions: 119H×330D mm 

Warranty: Two years European warranty

Regulatory approval: CE, FCC, ROHS

 : Note: EDOF, EDR, and DPQ are available on Windows OS only.

Technische Daten
  • Vergrößerung:
    700× - 900×
  • Auflösung:
    5,0 Megapixel
  • Bezeichnung:
    Mikroskop mit hoher Vergrößerung
  • Beleuchtung:
    8 LED Weiß
  • Cat. No.:
    IDCPAM73915MT8
  • Bildrate:
    15 fps bei 5,0 MP, YUY2 (USB 3,0) 45 fps bei 1,3 MP, YUY2 (USB 3,0) 15 fps bei 1,3 MP, YUY2 (USB 2,0)
  • Software:
    DinoCapture 2.0 (Windows), DinoXcope (Mac OS)
  • Gewicht:
    110 g
  • Sensor:
    CMOS