Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
Methyl nonadecanoate ≥99% Long chain fatty acid methyl ester

Methyl nonadecanoate ≥99% Long chain fatty acid methyl ester

Katalognr. ABCAAB143874-100
Anbieter:   Abcam
Methyl nonadecanoate ≥99% Long chain fatty acid methyl ester
Katalognr. ABCAAB143874-100
Anbieter:  Abcam
CAS Nummer:  
Methyl nonadecanoate ≥99% Long chain fatty acid methyl ester
CAS Nummer:  
Preis
SFr. 150.00
Einloggen für Verfügbarkeit
1 * 100 mg
Product Details & Documents

Long chain fatty acid methyl ester. Possible antitumor and anticlastogenic agent.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Synonyme:  Nonadecanoic acid methyll ester, Methyl Nonadecanoate
  • Molekulargewicht:312,5 g/mol
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.