AuftragsverfolgungKontakt
Ac-WEHD-AFC ≥97% | Avantor
Ac-WEHD-AFC ≥97%
Anbieter:   Abcam
Ac-WEHD-AFC ≥97%
Anbieter:  Abcam
Zertifikate
2 Varianten
Product Details & Documents
Ac-WEHD-AFC is a useful fluorogenic substrate for group I caspases (caspase-1, -4, and -5). Caspase activity can be quantified by fluorescent detection of free AFC that has an excitation maximum at 400 nm and emission maximum at 505 nm.
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Synonyme:  Caspase-1 substrate (fluorogenic), Caspase-5 substrate (fluorogenic)
  • Molekulargewicht:838,7 g/mol
Product Family Options
eral Biochemicals
Produktinformationen
Recommandations
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.