Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
Leptomycin B -Lösung 2,5% in Ethanol ≥95%, antifungal Inhibitor of CRM-1 mediated nuclear export. | Avantor

Leptomycin B -Lösung 2,5% in Ethanol ≥95%, antifungal Inhibitor of CRM-1 mediated nuclear export.

Anbieter:   Abcam
Leptomycin B -Lösung 2,5% in Ethanol ≥95%, antifungal Inhibitor of CRM-1 mediated nuclear export.
Anbieter:  Abcam
Zertifikate
Leptomycin B -Lösung 2,5% in Ethanol ≥95%, antifungal Inhibitor of CRM-1 mediated nuclear export.

2 Varianten

Product Details & Documents

Potent inhibitor of CRM-1-mediated (exportin-1) nuclear export. Displays antibiotic, antifungal, and antitumor properties and blocks the cell cycle at G₁ and G₂ phases. Potent inhibitor of nucleo-cytoplasmic translocation of Rev. Also shown to suppress HIV-1 replication in human monocytes (IC₅₀ = 600 pM).

Achtung! Bitte beachten: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Synonyme:  (2E,5S,6R,7S,9R,10E,12E,15R,16Z,18E)-17-Ethyl-6-hydroxy-3,5,7,9,11,15-hexamethyl-19-((2S,3S)-3-methyl-6-oxo-3,6-dihydro-2H-pyran-2-yl)-8-oxononadeca-2,10,12,16,18-pentaenoic acid
  • CAS Nummer:87081-35-4
  • Molekulargewicht:540,7 g/mol
Spezifikationen für diese Produktfamilie

Ergebnisse des Spezifikationstests

  • Constituents::
    97,5% Ethanol, 2,5% Leptomycin B
Product Family Options

Produktinformationen

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.