Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
Dihydroethidium fluoreszierender Farbstoff | Avantor

Dihydroethidium fluoreszierender Farbstoff

Katalognr. ABCAAB145360-100
Anbieter:   Abcam
Dihydroethidium fluoreszierender Farbstoff
Katalognr. ABCAAB145360-100
Anbieter:  Abcam
CAS Nummer:  104821-25-2
Dihydroethidium fluoreszierender Farbstoff
CAS Nummer:  104821-25-2
Preis
SFr. 454.00
Einloggen für Verfügbarkeit
1 * 100 mg
Product Details & Documents

Cell-permeable blue fluorescent dye. Redox indicator. Loses fluorescence on oxidation to ethidium. Detects superoxide radical in vivo.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Synonyme:  3,8-Diamino-5,6-dihydro-5-ethyl-6-phenylphenanthridine, Hydroethidine, DHE, 5-Ethyl-6-phenyl-5,6-dihydrophenanthridine-3,8-diamine, 2,7-Diamino-10-ethyl-9-phenyl-9,10-dihydrophenanthridine
  • CAS Nummer:104821-25-2
  • Molekulargewicht:315,4 g/mol
Technische Daten

Ergebnisse des Spezifikationstests

  • Constituents::
    100% Dihydroethidium
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.