Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
Margaric acid ≥99% Long chain saturated fatty acid

Margaric acid ≥99% Long chain saturated fatty acid

Katalognr. ABCAAB143871-1
Marketsource
Anbieter:   Abcam
Margaric acid ≥99% Long chain saturated fatty acid
Katalognr. ABCAAB143871-1
Anbieter:  Abcam
Marketsource
CAS Nummer:  506-12-7
Margaric acid ≥99% Long chain saturated fatty acid
CAS Nummer:  506-12-7
Preis
SFr. 340.00
Einloggen für Verfügbarkeit
1 * 1 g
Product Details & Documents

Long chain saturated fatty acid. Metabolite in glutamate and lipid metabolism in vivo. Blood-brain barrier permeable. Orally active.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Synonyme:  Heptadecanoic acid, Heptadecylic acid
  • CAS Nummer:506-12-7
  • Molekulargewicht:270,5 g/mol
Technische Daten

Ergebnisse des Spezifikationstests

  • Constituents::
    100% Heptadecanoic acid
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.