AC-55541 ≥98% protease-activated receptor 2 (PAR2) agonist
Über diesen Artikel
Potent, selective protease-activated receptor 2 (PAR2) agonist that displays no activity at other PAR subtypes or at over 30 other receptors involved in nociception and inflammation. Stimulates cell proliferation, PI hydrolysis and Ca²⁺ mobilization in vitro(pEC50 values are 6,7, 5,9 and 6,6 respectively) and exhibits pronociceptive activity in vivo.
Achtung! Bitte beachten: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
- CAS Nummer:916170-19-9
- Molekulargewicht:518,4 g/mol
2 Varianten
Product Details & Documents
Potent, selective protease-activated receptor 2 (PAR2) agonist that displays no activity at other PAR subtypes or at over 30 other receptors involved in nociception and inflammation. Stimulates cell proliferation, PI hydrolysis and Ca²⁺ mobilization in vitro(pEC50 values are 6,7, 5,9 and 6,6 respectively) and exhibits pronociceptive activity in vivo.
Achtung! Bitte beachten: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
- CAS Nummer:916170-19-9
- Molekulargewicht:518,4 g/mol
Spezifikationen für diese Produktfamilie
Ergebnisse des Spezifikationstests
- Constituents::100% AC 55541
Product Family Options
eral Biochemicals
Produktinformationen
Empfehlungen werden nur dann basierend auf Ihren Einkaufspräferenzen personalisiert, wenn Sie auf der Seite "Persönliche Daten" die Option „Mein Einkaufserlebnis verbessern“ aktiviert haben.
Andernfalls erhalten Sie allgemeine Empfehlungen..



