Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
Folic acid ≥96% Synthetic vitamin B9 | Avantor

Folic acid ≥96% Synthetic vitamin B9

Katalognr. ABCAAB143621-25
Anbieter:   Abcam
Folic acid ≥96% Synthetic vitamin B9
Katalognr. ABCAAB143621-25
Anbieter:  Abcam
CAS Nummer:  59-30-3
Folic acid ≥96% Synthetic vitamin B9
CAS Nummer:  59-30-3
Preis
SFr. 185.00
Einloggen für Verfügbarkeit
1 * 25 g
Product Details & Documents

Synthetic vitamin B9, Tetrahydrofolate precursor. Promotes DNA synthesis and cell division. Shows anti-inflammatory and chemopreventive effects *in vivo.* Orally active.

Achtung! Bitte beachten: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Synonyme:  Vitamin M, (S)-2-(4-(((2-Amino-4-oxo-3,4-dihydropteridin-6-yl)methyl)amino)benzamido)pentanedioic acid, Pteroyl-L-glutamic acid, PteGlu, Vitamin B9
  • CAS Nummer:59-30-3
  • Molekulargewicht:441,4 g/mol
Technische Daten

Ergebnisse des Spezifikationstests

  • Constituents::
    100% Folic acid