Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
Deoxythymidine triphosphate (dTTP) ≥98% nucleoside triphosphate

Deoxythymidine triphosphate (dTTP) ≥98% nucleoside triphosphate

Catalog # ABCAAB146540-50
Marketsource
Supplier:   Abcam
Deoxythymidine triphosphate (dTTP) ≥98% nucleoside triphosphate
Catalog # ABCAAB146540-50
Supplier:  Abcam
Marketsource
CAS Number:  18423-43-3
Deoxythymidine triphosphate (dTTP) ≥98% nucleoside triphosphate
CAS Number:  18423-43-3
Price
SFr. 235.00
Log in for availability
1 * 50 mg
Product Details & Documents

Nucleoside triphosphate used in many molecular biology applications such as DNA sequencing, PCR and other DNA polymerase based DNA synthesis and repair techniques.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Synonyms:  Thymidine-5'-triphosphoric acid, Deoxythymidine triphosphate, TPP, 2'-Deoxythymidine-5'-triphosphoric acid, dTTP
  • CAS Number:18423-43-3
  • Molecular Weight:548,12 g/mol
Specifications

Specification Test Results

  • Constituents::
    100% 2'-Deoxythymidine-5'-triphosphate [dTTP] trisodium
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.