Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
Cresol red Triarylmethane dye

Cresol red Triarylmethane dye

Marketsource
Supplier:   Abcam
Cresol red Triarylmethane dye
Supplier:  Abcam
Marketsource
Certificates

2 Options Available Below

Product Details & Documents

Triarylmethane dye commonly used with lead acetate for the detection of urease after polyacrylamide gel electrophoresis (PAGE). Also used as a pH indicator and as a tracking dye in DNA, RNA (agarose) and protein (polyacrylamide) electrophoresis. In agarose, Cresol Red runs with an apparent molecular size of approximately 125 bp DNA. Cresol Red does not inhibit Taq polymerase.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Synonyms:  o-Cresolsulphonphthalein, 3,3-Bis(4-hydroxy-3-methylphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide
  • CAS Number:1733-12-6
  • Molecular Weight:382,43 g/mol
Specifications for Product Family

Specification Test Results

  • Constituents::
    100% Cresol red
Product Family Options

Product Information

  • Availability
    Price
  • Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - SFr. 275.00
    SFr. 275.00
    1 * 25 g
       SFr. 275.00

    Specifications:
    Marketsource
    Marketsource
  • Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - SFr. 74.90
    SFr. 74.90
    1 * 5 g
       SFr. 74.90

    Specifications:
    Marketsource
    Marketsource
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.