Thymol blue pH-Indikator
Katalognr. ABCAAB146291-25
Anbieter: Abcam
Direkt zu
Product Details & Documents
pH indicator with visual transition intervals of pH 1,2 (red) to pH 2,8 (yellow), and pH 8,0 (yellow) to pH 9,2 (blue).
: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Synonyme: Thymolsulphonphthalein, 3,3-Bis(4-hydroxy-5-isopropyl-2-methylphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide
- CAS Nummer:76-61-9
- Molekulargewicht:466,6 g/mol
Technische Daten
Ergebnisse des Spezifikationstests
- Constituents::100% Thymol blue



