Monosodium urate Pro-inflammatory agent
NLRP3/NALP3 inflammasome activator. Stimulates caspase-1 activating NLRP3/NALP3 inflammasome, results in active IL-1β and IL-18 production. Active in vitroand in vivo.
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
- :1198-77-2
- :190,09 g/mol
- Constituents::100% Monosodium urate
- Cat. No.:ABCAAB144305-2