Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
Nile Red ≥95% lipophilic stain

Nile Red ≥95% lipophilic stain

Katalognr. ABCAAB219403-25
Marketsource
Anbieter:   Abcam
Nile Red ≥95% lipophilic stain
Katalognr. ABCAAB219403-25
Anbieter:  Abcam
Marketsource
CAS Nummer:  7385-67-3
Nile Red ≥95% lipophilic stain
CAS Nummer:  7385-67-3
Preis
SFr. 170.00
Einloggen für Verfügbarkeit
1 * 25 mg
Product Details & Documents

Photostable, lipophilic stain that strongly fluoresces in hydrophobic (lipid-rich) environments, but has minimal fluorescence in aqueous media. Commonly used for the detection of intracellular lipid droplets by fluorescence microscopy and flow cytofluorometry.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Synonyme:  9-(Diethylamino)-5H-benzo[a]phenoxazin-5-one
  • CAS Nummer:7385-67-3
  • Molekulargewicht:318,4 g/mol
Technische Daten

Ergebnisse des Spezifikationstests

  • Constituents::
    100% Nile Red
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.