Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
β-Sitosterol ≥98% Antitumor agent | Avantor

β-Sitosterol ≥98% Antitumor agent

Anbieter:   Abcam
β-Sitosterol ≥98% Antitumor agent
Anbieter:  Abcam
Zertifikate

3 Varianten

Product Details & Documents

Plant sterol. Various pharmacological effects. Antitumor agent. Activates caspase 3 and induces G2/M arrest and DNA fragmentation. Increases α-tubulin levels and promotes microtubule polymerization. Inhibits dietary cholesterol absorption in vivo. Orally active.

Achtung! Bitte beachten: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Synonyme:  α-Dihydrofucosterol, β-Sitosterol, 5-Stigmasten-3β-ol, 22,23-Dihydrostigmasterol, 24α-Ethylcholesterol
  • CAS Nummer:83-46-5
  • Molekulargewicht:414,7 g/mol
Spezifikationen für diese Produktfamilie

Ergebnisse des Spezifikationstests

  • Constituents::
    98% ß-Sitosterol
Product Family Options

Produktinformationen

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.