Lignoceric acid ≥99% Long chain saturated fatty acid
Product Details & Documents
Long chain saturated fatty acid. Component of cerebrosides and sphingomyelin. Acyl-CoA ligase substrate. Energy source in vivo. Orally active.
Achtung! Bitte beachten: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
- CAS Nummer:557-59-5
- Molekulargewicht:368,6 g/mol
Technische Daten
Ergebnisse des Spezifikationstests
- Constituents::100% Tetracosanoic acid
Empfehlungen werden nur dann basierend auf Ihren Einkaufspräferenzen personalisiert, wenn Sie auf der Seite "Persönliche Daten" die Option „Mein Einkaufserlebnis verbessern“ aktiviert haben.
Andernfalls erhalten Sie allgemeine Empfehlungen..



