Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
PRC Pathway Modulator Set Abcam | Avantor

PRC Pathway Modulator Set Abcam

PRC Pathway Modulator Set
Katalognr. ABCAAB284177-1
Anbieter:   Abcam
PRC Pathway Modulator Set Abcam
Katalognr. ABCAAB284177-1
Anbieter:  Abcam
CAS Nummer:  
PRC Pathway Modulator Set Abcam

PRC Pathway Modulator Set

CAS Nummer:  
Preis
SFr. 459.00
Einloggen für Verfügbarkeit
1 * 1 KIT
Product Details & Documents

PRC Pathway Modulator Set (ab284177) is part of the multiplex kits range. Abcam offers high-quality biological reagents and tools including antibodies, proteins, assays, cell lines and lysates.

Achtung! Bitte beachten: All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Technische Daten

Technische Daten

  • Bezeichnung:
    PRC Pathway Modulator Set
  • Cat. No.:
    ABCAAB284177-1
  • Lagertemperatur:
    Store at -20°C.

Technische Daten