Antigen Retrieval Buffer (100X Citrate Buffer), Abcam
Über diesen Artikel
Antigen retrieval 100X citrate buffer (pH 5.1 - 5.5) is a special solution that enables rehydration and antigen retrieval to be performed in formalin-fixed, paraffin-embedded tissue sections with one step. - Cited in over 25 publications - Contains citrate buffer, 1.5% 2-butoxyethanol, 1% hydrochloric acid - Optimized for citrate buffer-treated antibodies - Contains detergent for paraffin emulsification
- pH: 5.1 - 5.5 Constituents: Citrate Buffer, 1.5% 2-butoxyethanol, 1% Hydrochloric acid
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
2 Varianten
Product Details & Documents
Antigen retrieval 100X citrate buffer (pH 5.1 - 5.5) is a special solution that enables rehydration and antigen retrieval to be performed in formalin-fixed, paraffin-embedded tissue sections with one step. - Cited in over 25 publications - Contains citrate buffer, 1.5% 2-butoxyethanol, 1% hydrochloric acid - Optimized for citrate buffer-treated antibodies - Contains detergent for paraffin emulsification
- pH: 5.1 - 5.5 Constituents: Citrate Buffer, 1.5% 2-butoxyethanol, 1% Hydrochloric acid
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Spezifikationen für diese Produktfamilie
Technische Daten
Technische Daten
- Catalog No:
- ABCAAB93678-125
- ABCAAB93678-250
- Lagertemperatur:
- Store at Ambient.
- Store at Ambient.
Technische Daten
Product Family Options
ondary Antibodies
Produktinformationen
- BezeichnungVerfügbarkeitPreis
- Spezifikationen:Marketsource
Mehr
Specifications
Cat. No.ABCAAB93678-125LagertemperaturStore at Ambient.Marketsource
logical Buffers
Produktinformationen
- BezeichnungVerfügbarkeitPreis
- Spezifikationen:
Mehr
Specifications
Cat. No.ABCAAB93678-250LagertemperaturStore at Ambient.
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



