Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
Hydrogen Peroxide Blocking Reagent, Abcam

Hydrogen Peroxide Blocking Reagent, Abcam

Anbieter:   Abcam
Hydrogen Peroxide Blocking Reagent, Abcam
Anbieter:  Abcam
Zertifikate

2 Varianten

Product Details & Documents

Hydrogen peroxide blocking reagent is used to block endogenous peroxidase activity in IHC applications, enhancing signal detection in HRP / peroxidase-based systems. - Cited in over 30 publications - Endogenous peroxidase block – prevents non-specific background staining in IHC - HRP-based detection – ideal for use with HRP/peroxidase detection systems - Concentration – contains 0.3% hydrogen peroxide

  • Preservative: 0.1% Sodium azide Constituents: 0.3% Hydrogen peroxide

Caution: All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Spezifikationen für diese Produktfamilie

Technische Daten

Technische Daten

  • Catalog No:
  • ABCAAB64218-125
  • ABCAAB64218-60
  • Lagertemperatur:
  • Store at +4°C.
  • Store at +4°C.

Technische Daten

Product Family Options

Produktinformationen

  • Bezeichnung
    Verfügbarkeit
    Preis
  • Hydrogen Peroxide Blocking Reagent
    Einloggen für Verfügbarkeit
    Einloggen für Verfügbarkeit
    1 * 125 mL
    1 * 125 mL - SFr. 195.00
    SFr. 195.00
    1 * 125 mL
       SFr. 195.00

    Spezifikationen:
    Marketsource
    Mehr
    Specifications
    Cat. No.
    ABCAAB64218-125
    Lagertemperatur
    Store at +4°C.
    Marketsource
  • Hydrogen Peroxide Blocking Reagent
    Einloggen für Verfügbarkeit
    Einloggen für Verfügbarkeit
    1 * 60 mL
    1 * 60 mL - SFr. 130.00
    SFr. 130.00
    1 * 60 mL
       SFr. 130.00

    Spezifikationen:
    Mehr
    Specifications
    Cat. No.
    ABCAAB64218-60
    Lagertemperatur
    Store at +4°C.
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.