AuftragsverfolgungKontakt
G-Biosciences Hybridization Buffer (Hybridization Neutralizing Solution) | Avantor
G-Biosciences Hybridization Buffer (Hybridization Neutralizing Solution)
G-Biosciences Hybridization Buffer (Hybridization Neutralizing Solution)
Anbieter:  G-Biosciences
Zertifikate
G-Biosciences Hybridization Buffer (Hybridization Neutralizing Solution)
G-Biosciences Hybridization Buffer (Hybridization Neutralizing Solution)
2 Varianten
Product Details & Documents
Ready-to-Use: Convenient, time-saving formulation ideal for consistent experimental reproducibility.
  • 1 M Tris·HCl (pH 7,4)
  • 1,5 M sodium chloride
Used to neutralize membrane-bound samples and stabilize probe-target duplexes. The Tris buffer maintains physiological pH, while the high salt concentration supports nucleic acid duplex stability and reduces nonspecific binding.
Dokumente
Product Family Options
Produktinformationen
  • Bezeichnung
    VE
    Verfügbarkeit
    Preis
  • Hybridization neutralizing solution (1 M Tris.HCl (pH 7,4), 1,5 M NaCl)
    1 L
    Einloggen für Verfügbarkeit
    Einloggen für Verfügbarkeit
    1 * 1 L
    1 * 1 L - SFr. 90.60
    SFr. 90.60
    1 * 1 L
       SFr. 90.60
  • Hybridization neutralizing solution (1 M Tris.HCl (pH 7,4), 1,5 M NaCl)
    1 gal.
    Einloggen für Verfügbarkeit
    Einloggen für Verfügbarkeit
    1 * 4 L
    1 * 4 L - SFr. 173.00
    SFr. 173.00
    1 * 4 L
       SFr. 173.00