CRISPRclean® Stranded Total RNA Prep
Katalognr. BIOONOVA-5229950
Anbieter: BIOO SCIENTIFIC
CAS-Nummer:
Technische Daten
- Bezeichnung:CRISPRclean® stranded total RNA prep
Technische Daten
Über diesen Artikel
The CRISPRclean® stranded total RNA prep with rRNA depletion (HMR) is a total RNA library prep leveraging a CRISPR-powered workflow facilitating the detection of lower expressing and biologically relevant transcripts from complex samples.
- Efficient multi-species depletion using CRISPR-Cas9 technology in a single tube Enhances detection of biologically relevant, low-expressing transcripts Optimized library prep and depletion workflow ensures high-quality transcript representation Achieve consistent full-length, uniform transcript coverage with high library complexity Complete workflows in just one day: 7,5 hours assay with 3 hours of hands-on time
Specific, unbiased rRNA depletion and library prep for RNA-Seq. In RNA samples, up to 90% consists of ribosomal RNA. Failing to remove these abundant rRNA sequences during transcriptome sequencing reduces sequencing capacity and hinders the detection of biologically relevant, low-expressing transcripts. Discover how CRISPRclean® enhances research by efficiently removing rRNA.
Certifications: Manufactured in ISO 9001 facility.
Caution: For research use only. Not for use in diagnostic procedures.