Specifications
- Schuhgröße:40
- Farbe:Schwarz
Specifications
About this item
Slip resistance on ceramic, coated with SLS solution and on steel, coated with glycerol - slip resistance on ceramic substrate, covered with SLS solution and on steel substrate, covered with glycerol.
Unisex footwear
Occupational footwear
Increased durability of the upper
Antibacterial lining
Upper material: Grain leather
Lining material: Fabric
Lining material (features): Antibacterial lining
Insole: Fabric
Insole (features): Antibacterial insole, orthopedic insoles used, replaceable insole
Fastener type: Shoelaces
Antistatic footwear - antistatic properties
Energy absorption in the heel area - energy absorption in the heel area
The sole resistant to diesel fuel - sole resistance to diesel fuel
Water permeability and water absorption of the face material - permeability and water absorption of the upper material
Protection class: O2 - Basic requirements plus: Closed seat region (fully enclosed heel). Anti-static properties. Energy absorption of seat region. Plus, water penetration and absorption resistance.
EN ISO 20347:2012; 2102186 02 86.