Tork T4 Conventional Toilet Paper Roll, White, Universal
Catalog # SCAA472165
Supplier: Essity
New Product
Specifications
- Verpackt:8 packs, 6 rolls each. 200 sheets each.
- Bezeichnung:Universal conventional toilet paper roll white, compatible with T4 dispensers
- L×B:23800×94 mm
Specifications
About this item
Tork universal traditional roll toilet paper meets essential needs while offering a quality product at a low price for low-traffic washrooms.
Maximise cost-efficiency with this universal quality bulk toilet paper
- Meet the basic needs of low-traffic washrooms
-
Make more sustainable choices, the inner packaging contains a minimum of 30% post-consumer recycled plasticBoost hygiene by using these commercial toilet paper rolls with a Tork T4 dispenser, which has a neat closed design
Universal quality
Sheet length 119 mm
Core inner diameter 40 mm
Use these toilet paper rolls with the closed design of the Tork T4 dispenser for increased washroom hygiene.