Tork T4 Premium Extra Soft Conventional Toilet Paper Roll, White
Référence SCAA110406
Fournisseur: Essity
New Product
Spécifications
- Verpackt:7 packs, 6 rolls each. 150 sheets each, 4 ply.
- Bezeichnung:Premium conventional toilet paper roll, refill, white, compatible with Tork T4 dispensers
- L×B:18750×117 mm
Spécifications
À propos de cet article
Provide the guests with the comfort of being at home, away from home, with the Premium Tork conventional toilet roll. This extra-soft toilet paper offers a luxurious look and feel with superior performance for upmarket washrooms in hotels or office buildings.
Enhance the washroom decor with the attractive design of this bulk toilet paper
-
Carry rolls and dispose of packaging with ease due to the Tork Easy Handling® plastic packaging
Core inner diameter 42 mm
Enhance the superior feel with this bulk toilet paper's luxurious 4-ply construction and impressive performance. Plus, make life simpler for staff with the Tork Easy Handling® plastic packaging. It allows for easier roll carrying and packaging disposal.
Certifications:
FSC MIX Credit | SA-COC-008266. EU Ecolabel | SE/004/001.