Human Recombinant PGRPS (from E. coli)
Product Details & Documents
Recombinant Human PGRPS protein - BSA and Azide free is a Human Full Length protein, in the 22 to 196 aa range, expressed in Escherichia coli, with >90% purity, <1 EU/µg endotoxin level and suitable for SDS-PAGE, ELISA, WB, MS.
Achtung! Bitte beachten: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Technische Daten
Technische Daten
- Konjugation:Unconjugated
- Form:Lyophilised
- Transporttemperatur:Dry Ice
- Validierte Anwendungen:ELISA, MS, SDS-PAGE, WB
- Purity:>90%
- Spezies:Human
- Biologische Aktivität:Standard
- Zusammensetzung:Constituents: 0,41% Sodium acetate
- Cat. No.:ABCAAB175469-100
- Ursprung:E. coli
- Endotoxingehalt:<1 EU/µg
- Tierfrei:N
- Protein/Peptid-Typ:Rekombinant
- Lagerbedingungen:Store at −80 °C
- Protein/Peptid-Name:PGRPS
- Proteinsynonym:PGLYRP|PGRP|TNFSF3L|SBBI68|UNQ639/PRO1269|PGLYRP1|Peptidoglycan recognition protein 1|Peptidoglycan recognition protein short|PGRP-S
Technische Daten
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



