Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
Human Recombinant RPC8 (from E. coli) | Avantor

Human Recombinant RPC8 (from E. coli)

Katalognr. ABCAAB134603-100
Marketsource
Anbieter:   Abcam
Human Recombinant RPC8 (from E. coli)
Katalognr. ABCAAB134603-100
Anbieter:  Abcam
Marketsource
CAS Nummer:  
Human Recombinant RPC8 (from E. coli)
Human Recombinant RPC8 (from E. coli)
CAS Nummer:  
Preis
SFr. 529.00
Einloggen für Verfügbarkeit
1 * 100 µG
Product Details & Documents

Recombinant human RPC8 protein is a Human Full Length protein, in the 1 to 204 aa range, expressed in Escherichia coli, with >90% purity and suitable for SDS-PAGE, MS.

Achtung! Bitte beachten: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Technische Daten

Technische Daten

  • Konjugation:
    Unconjugated
  • Form:
    Liquid
  • Transporttemperatur:
    20
  • Validierte Anwendungen:
    MS, SDS-PAGE
  • Purity:
    >90%
  • Spezies:
    Human
  • Konzentration:
    0,5 mg/ml
  • Biologische Aktivität:
    Standard
  • Zusammensetzung:
    pH: 8, Constituents: 10% Glycerol (glycerin, glycerine), 0,58% Sodium chloride, 0,32% Tris HCl, 0,02% (R*,R*)-1,4-Dimercaptobutan-2,3-diol
  • Cat. No.:
    ABCAAB134603-100
  • Ursprung:
    E. coli
  • Tierfrei:
    N
  • Protein/Peptid-Typ:
    Rekombinant
  • Lagerbedingungen:
    Store at −20 °C
  • Protein/Peptid-Name:
    RPC8
  • Proteinsynonym:
    KIAA1665|RPC8|POLR3H|DNA-directed RNA polymerase III subunit RPC8|RNA polymerase III subunit C8|DNA-directed RNA polymerase III subunit H|RNA polymerase III subunit 22.9 kDa subunit|RPC22.9

Technische Daten