Human Recombinant IL-1 beta (from E. coli)
Über diesen Artikel
Recombinant human IL-1 beta protein (Active) is a Human Full Length protein, in the 117 to 269 aa range, expressed in Escherichia coli, with >98% purity, <1 EU/µg endotoxin level and suitable for ELISA, FuncS, HPLC, SDS-PAGE.
: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
2 Varianten
Product Details & Documents
Recombinant human IL-1 beta protein (Active) is a Human Full Length protein, in the 117 to 269 aa range, expressed in Escherichia coli, with >98% purity, <1 EU/µg endotoxin level and suitable for ELISA, FuncS, HPLC, SDS-PAGE.
: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Spezifikationen für diese Produktfamilie
Technische Daten
Technische Daten
- Catalog No:
- ABCAAB9617-10
- ABCAAB9617-2
- Protein/Peptid-Name:
- IL-1 beta
- IL-1 beta
- Proteinsynonym:
- IL1F2|IL1B|Interleukin-1 beta|Catabolin
- IL1F2|IL1B|Interleukin-1 beta|Catabolin
- Protein/Peptid-Typ:
- Rekombinant
- Rekombinant
- Spezies:
- Human
- Human
- Ursprung:
- E. coli
- E. coli
- Konjugation:
- Unconjugated
- Unconjugated
- Tierfrei:
- N
- N
- Endotoxingehalt:
- <1 EU/µg
- <1 EU/µg
- Biologische Aktivität:
- Bioactive
- Bioactive
- Purity:
- >98%
- >98%
- Form:
- Lyophilised
- Lyophilised
- Zusammensetzung:
- Constituents: 0,3% Sodium chloride, 0,16% Sodium phosphate
- Constituents: 0,3% Sodium chloride, 0,16% Sodium phosphate
- Lagerbedingungen:
- Store at –20 °C
- Store at –20 °C
- Transporttemperatur:
- 20
- 20
- Validierte Anwendungen:
- ELISA, FuncS, HPLC, SDS-PAGE
- ELISA, FuncS, HPLC, SDS-PAGE
Technische Daten
Product Family Options
Produktinformationen
- VerfügbarkeitPreis
- Spezifikationen:
Mehr
Specifications
Cat. No.ABCAAB9617-10Protein/Peptid-NameIL-1 betaProteinsynonymIL1F2|IL1B|Interleukin-1 beta|CatabolinProtein/Peptid-TypRekombinantSpeziesHumanUrsprungE. coliKonjugationUnconjugatedTierfreiNEndotoxingehalt<1 EU/µgBiologische AktivitätBioactivePurity>98%FormLyophilisedZusammensetzungConstituents: 0,3% Sodium chloride, 0,16% Sodium phosphateLagerbedingungenStore at –20 °CTransporttemperatur20Validierte AnwendungenELISA, FuncS, HPLC, SDS-PAGE - Spezifikationen:
Mehr
Specifications
Cat. No.ABCAAB9617-2Protein/Peptid-NameIL-1 betaProteinsynonymIL1F2|IL1B|Interleukin-1 beta|CatabolinProtein/Peptid-TypRekombinantSpeziesHumanUrsprungE. coliKonjugationUnconjugatedTierfreiNEndotoxingehalt<1 EU/µgBiologische AktivitätBioactivePurity>98%FormLyophilisedZusammensetzungConstituents: 0,3% Sodium chloride, 0,16% Sodium phosphateLagerbedingungenStore at –20 °CTransporttemperatur20Validierte AnwendungenELISA, FuncS, HPLC, SDS-PAGE



